Methyl-3-(2,6-dichlorophenyl)-5-methylisoxazole-4-carboxylate
Supplier: Apollo Scientific
OR29819-10G
OR29819-1G
OR29819-5G
APOSOR29819-10GEA
316
EUR
APOSOR29819-10G
APOSOR29819-1G
APOSOR29819-5G
Methyl-3-(2,6-dichlorophenyl)-5-methylisoxazole-4-carboxylate
Methyl-3-(2,6-dichlorophenyl)-5-methylisoxazole-4-carboxylate
Formula:
C₁₂H₉Cl₂NO₃ MW: 286,12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00115005 CAS Number: 4402-83-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...