Oxazolo[4,5-b]pyridin-2(3H)-one

Supplier: Apollo Scientific

Synonyms: 2,3-Dihydropyrido[2,3-d]oxazol-2-one

OR29854-1G OR29854-5G
APOSOR29854-1GEA 49.5 EUR
APOSOR29854-1G APOSOR29854-5G
Oxazolo[4,5-b]pyridin-2(3H)-one
Oxazolo[4,5-b]pyridin-2(3H)-one
Formula: C₆H₄N₂O₂
MW: 136,11 g/mol
Melting Pt: 210…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00204215
CAS Number: 60832-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR