2-Chloroisonicotinoyl chloride 95%
Supplier: Apollo Scientific
|
Danger
|
OR29882-1G
OR29882-5G
OR29882-10G
OR29882-250MG
APOSOR29882-1GEA
98.8
EUR
APOSOR29882-1G
APOSOR29882-5G
APOSOR29882-10G
APOSOR29882-250MG
2-Chloroisonicotinoyl chloride 95%
2-Chloroisonicotinoyl chloride
Formula:
C₆H₃Cl₂NO Boiling Pt: 144 °C Density: 1,418 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00204232 CAS Number: 65287-34-5 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...