Methyl-3-methylsalicylate 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Methyl 2-hydroxy-3-methylbenzoate
OR29986-1G
OR29986-5G
APOSOR29986-1GEA
21.9
EUR
APOSOR29986-1G
APOSOR29986-5G
Methyl-3-methylsalicylate 97%
Methyl 3-methylsalicylate
Formula:
C₉H₁₀O₃ MW: 166,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00020051 CAS Number: 23287-26-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...