5-Bromo-1-chloroisoquinoline
Supplier: Apollo Scientific
|
Warning
|
OR300068-1G
OR300068-250MG
APOSOR300068-1GEA
145
EUR
APOSOR300068-1G
APOSOR300068-250MG
5-Bromo-1-chloroisoquinoline
5-Bromo-1-chloroisoquinoline
Formula:
C₉H₅BrClN MW: 242,4997 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08703278 CAS Number: 34551-41-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...