4-Bromo-1-chloroisoquinoline 95%
Supplier: Apollo Scientific
|
Warning
|
OR300096-1G
OR300096-10G
OR300096-5G
APOSOR300096-1GEA
107
EUR
APOSOR300096-1G
APOSOR300096-10G
APOSOR300096-5G
4-Bromo-1-chloroisoquinoline 95%
4-Bromo-1-chloroisoquinoline
Formula:
C₉H₅BrClN MW: 242,4997 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00234469 CAS Number: 66728-98-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...