2-Aminobenzo[d]thiazole-5-carbonitrile
Supplier: Apollo Scientific
|
Warning
|
OR300283-1G
OR300283-250MG
APOSOR300283-1GEA
394
EUR
APOSOR300283-1G
APOSOR300283-250MG
2-Aminobenzo[d]thiazole-5-carbonitrile
2-Aminobenzo[d]thiazole-5-carbonitrile
Formula:
C₈H₅N₃S MW: 175,21 g/mol Storage Temperature: Ambient |
CAS Number:
105314-08-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...