Ethyl-4-chlorobenzoylformate 97%
Supplier: Apollo Scientific
|
Warning
|
OR30031-10G
OR30031-25G
OR30031-5G
APOSOR30031-10GEA
84.1
EUR
APOSOR30031-10G
APOSOR30031-25G
APOSOR30031-5G
Ethyl-4-chlorobenzoylformate 97%
Ethyl 4-chlorobenzoylformate
Formula:
C₁₀H₉ClO₃ Boiling Pt: 85 °C/0,1mmHg Storage Temperature: Ambient |
MDL Number:
MFCD01319615 CAS Number: 34966-48-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...