5-Iodo-1,2,3-trimethoxybenzene

Supplier: Apollo Scientific
Warning

OR3004-1G OR3004-5G
APOSOR3004-1GEA 64 EUR
APOSOR3004-1G APOSOR3004-5G
5-Iodo-1,2,3-trimethoxybenzene
5-Iodo-1,2,3-trimethoxybenzene
Formula: C₉H₁₁IO₃
MW: 294,09 g/mol
Melting Pt: 85…87 °C
Storage Temperature: Ambient
MDL Number: MFCD01318153
CAS Number: 25245-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR