Ethyl-3-(ethoxycarbonylmethylamino)propionate

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 3-((2-ethoxy-2-oxoethyl)amino)propanoate

OR300513-1G OR300513-10G OR300513-5G OR300513-250MG OR300513-500MG OR300513-25G
APOSOR300513-1GEA 98.9 EUR
APOSOR300513-1G APOSOR300513-10G APOSOR300513-5G APOSOR300513-250MG APOSOR300513-500MG APOSOR300513-25G
Ethyl-3-(ethoxycarbonylmethylamino)propionate
Ethyl-3-(ethoxycarbonylmethylamino)propionate
Formula: C₉H₁₇NO₄
MW: 203,24 g/mol
Boiling Pt: 117 °C/0,5mmHg
Density: 1,0533 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD08729298
CAS Number: 3783-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR