4-(3-Chloropropyl)morpholine

Supplier: Apollo Scientific
Warning

Synonyms: 3-Morpholinopropyl Chloride

OR300520-100G OR300520-25G
APOSOR300520-100GEA 55.1 EUR
APOSOR300520-100G APOSOR300520-25G
4-(3-Chloropropyl)morpholine
4-(3-Chloropropyl)morpholine
Formula: C₇H₁₄ClNO
MW: 163,65 g/mol
Boiling Pt: 115 °C/25mmHg
Density: 1,08 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00039714
CAS Number: 7357-67-7
EINECS: 461-510-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR