Ethyl-5-pyridin-4-yl-1,2,4-oxadiazole-3-carboxylate
Supplier: Apollo Scientific
OR300589-1G
OR300589-250MG
OR300589-500MG
OR300589-5G
APOSOR300589-1GEA
393
EUR
APOSOR300589-1G
APOSOR300589-250MG
APOSOR300589-500MG
APOSOR300589-5G
Ethyl-5-pyridin-4-yl-1,2,4-oxadiazole-3-carboxylate
Ethyl-5-pyridin-4-yl-1,2,4-oxadiazole-3-carboxylate
Formula:
C₁₀H₉N₃O₃ MW: 219,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09991789 CAS Number: 151097-45-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...