4-Chloropyridine-2-carbonitrile 98%
Supplier: Apollo Scientific
|
Danger
|
OR300669-1G
OR300669-5G
OR300669-25G
APOSOR300669-1GEA
21.9
EUR
APOSOR300669-1G
APOSOR300669-5G
APOSOR300669-25G
4-Chloropyridine-2-carbonitrile 98%
4-Chloropyridine-2-carbonitrile
Formula:
C₆H₃ClN₂ MW: 138,5500 g/mol Melting Pt: 81…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD06009833 CAS Number: 19235-89-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...