3-Iodo-o-xylene
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1,2-Dimethyl-3-iodobenzene, 2,3-Dimethyl-1-iodobenzene, 1-Iodo-2,3-dimethylbenzene, 2,3-Dimethyliodobenzene
OR30083-25G
OR30083-5G
APOSOR30083-25GEA
72.7
EUR
APOSOR30083-25G
APOSOR30083-5G
3-Iodo-o-xylene
3-Iodo-o-xylene
Formula:
C₈H₉I MW: 232,06 g/mol Boiling Pt: 98…100 °C/15mmHg Density: 1,64 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00041437 CAS Number: 31599-60-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...