3-Bromo-2-methylanisole
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Bromo-3-methoxy-2-methylbenzene
OR301007-1G
OR301007-5G
APOSOR301007-1GEA
56.8
EUR
APOSOR301007-1G
APOSOR301007-5G
3-Bromo-2-methylanisole
3-Bromo-2-methylanisole
Formula:
C₈H₉BrO MW: 201,0605 g/mol Boiling Pt: 115…116 °C/20mmHg Storage Temperature: Refrigerator |
MDL Number:
MFCD11100991 CAS Number: 31804-36-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...