4-Cbz-aminopiperidine
Supplier: Apollo Scientific
OR301035-1G
OR301035-5G
OR301035-10G
APOSOR301035-1GEA
78.6
EUR
APOSOR301035-1G
APOSOR301035-5G
APOSOR301035-10G
4-Cbz-aminopiperidine
4-Cbz-aminopiperidine
Formula:
C₁₃H₁₈N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD04038571 CAS Number: 182223-54-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...