4-Iodo-2-nitroaniline 98%
Supplier: Apollo Scientific
|
Warning
|
OR301096-1G
OR301096-5G
OR301096-10G
APOSOR301096-1GEA
21.9
EUR
APOSOR301096-1G
APOSOR301096-5G
APOSOR301096-10G
4-Iodo-2-nitroaniline 98%
4-Iodo-2-nitroaniline
Formula:
C₆H₅IN₂O₂ MW: 264,02 g/mol Melting Pt: 120…123 °C Storage Temperature: Ambient |
MDL Number:
MFCD06808526 CAS Number: 20691-72-9 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...