5-Bromo-1-methyl-1H-pyrazole
Supplier: Apollo Scientific
|
Warning
|
OR301097-1G
OR301097-100MG
OR301097-5G
APOSOR301097-1GEA
79.9
EUR
APOSOR301097-1G
APOSOR301097-100MG
APOSOR301097-5G
5-Bromo-1-methyl-1H-pyrazole
5-Bromo-1-methyl-1H-pyrazole
Formula:
C₄H₅BrN₂ Storage Temperature: Ambient |
MDL Number:
MFCD11656572 CAS Number: 361476-01-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...