Methyl-2-(chloromethyl)thiazole-4-carboxylate 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
Methyl 2-(chloromethyl)thiazole-4-carboxylate
OR301119-1G
OR301119-250MG
APOSOR301119-1GEA
37.7
EUR
APOSOR301119-1G
APOSOR301119-250MG
Methyl-2-(chloromethyl)thiazole-4-carboxylate 97%
Methyl-2-(chloromethyl)thiazole-4-carboxylate
Formula:
C₆H₆ClNO₂S MW: 191,6400 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08669440 CAS Number: 321371-29-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...