3-Methyl-1H-pyrazole 99%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Methylpyrazole

OR301129-100G OR301129-25G OR301129-500G
APOSOR301129-100GEA 55.1 EUR
APOSOR301129-100G APOSOR301129-25G APOSOR301129-500G
3-Methyl-1H-pyrazole 99%
3-Methyl-1H-pyrazole
Formula: C₄H₆N₂
MW: 82,11 g/mol
Boiling Pt: 203…205 °C
Density: 1,02 g/cm³ (25 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00005240
CAS Number: 1453-58-3
EINECS: 215-925-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR