3,4-Dimethylbenzaldehyde
Supplier: Apollo Scientific
|
Warning
|
OR30113-100G
OR30113-25G
APOSOR30113-100GEA
24.8
EUR
APOSOR30113-100G
APOSOR30113-25G
3,4-Dimethylbenzaldehyde
3,4-Dimethylbenzaldehyde
Formula:
C₉H₁₀O MW: 134,18 g/mol Boiling Pt: 102…104 °C/15mmHg Melting Pt: 223…226 °C Density: 1,012 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00016612 CAS Number: 5973-71-7 EINECS: 227-770-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...