3,4-Dichloro-N-methylaniline

Supplier: Apollo Scientific

Synonyms: N1-Methyl-3,4-dichloroaniline

OR30114-10G OR30114-1G OR30114-5G
APOSOR30114-10GEA 0 EUR
APOSOR30114-10G APOSOR30114-1G APOSOR30114-5G
3,4-Dichloro-N-methylaniline
3,4-Dichloro-N-methylaniline
Formula: C₇H₇Cl₂N
MW: 176,04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00052850
CAS Number: 40750-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR