5-Bromo-3-chloropyridin-2-amine 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Amino-5-bromo-3-chloropyridine

OR301145-1G OR301145-5G
APOSOR301145-1GEA 69.8 EUR
APOSOR301145-1G APOSOR301145-5G
5-Bromo-3-chloropyridin-2-amine 98%
5-Bromo-3-chloropyridin-2-amine
Formula: C₅H₄BrClN₂
MW: 207,46 g/mol
Melting Pt: 94…96 °C
Storage Temperature: Ambient
MDL Number: MFCD11110443
CAS Number: 38185-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR