2-Acetylbenzonitrile 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Acetylbenzenecarbonitrile

OR301156-1G OR301156-5G OR301156-10G
APOSOR301156-1GEA 42.2 EUR
APOSOR301156-1G APOSOR301156-5G APOSOR301156-10G
2-Acetylbenzonitrile 97%
2-Acetylbenzonitrile
Formula: C₉H₇NO
MW: 145,1600 g/mol
Boiling Pt: 148 °C/12mmHg
Melting Pt: 46…49 °C
Storage Temperature: Ambient
MDL Number: MFCD00272571
CAS Number: 91054-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR