Isonicotinonitrile
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Pyridinecarbonitrile, Pyridine-4-carbonitrile, 4-Cyanopyridine
OR301182-100G
OR301182-500G
OR301182-1KG
APOSOR301182-100GEA
29
EUR
APOSOR301182-100G
APOSOR301182-500G
APOSOR301182-1KG
Isonicotinonitrile
Isonicotinonitrile
Formula:
C₆H₄N₂ Melting Pt: 76…79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006417 CAS Number: 100-48-1 EINECS: 202-856-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...