Ethyl-2-methylquinoline-3-carboxylate

Supplier: Apollo Scientific
Danger

Synonyms: Ethyl 2-methylquinoline-3-carboxylate, 2-Methyl-quinoline-3-carboxylic acid ethyl ester

OR301187-1G OR301187-250MG
APOSOR301187-1GEA 81.2 EUR
APOSOR301187-1G APOSOR301187-250MG
Ethyl-2-methylquinoline-3-carboxylate
Ethyl-2-methylquinoline-3-carboxylate
Formula: C₁₃H₁₃NO₂
MW: 215,25 g/mol
Melting Pt: 70…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00195614
CAS Number: 15785-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR