2-Methyl-5-pyrimidineboronic acid 96%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(2-Methylpyrimidin-5-yl)boronic acid, 2-Methylpyrimidine-5-boronic acid
OR301199-1G
OR301199-250MG
OR301199-5G
APOSOR301199-1GEA
108
EUR
APOSOR301199-1G
APOSOR301199-250MG
APOSOR301199-5G
2-Methyl-5-pyrimidineboronic acid 96%
2-Methyl-5-pyrimidineboronic acid
Formula:
C₅H₇BN₂O₂ MW: 137,9323 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD07375144 CAS Number: 1034924-06-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...