5-Bromopyrimidine-2-carboxylic acid 97%

Supplier: Apollo Scientific
Warning

OR301240-25G OR301240-5G
APOSOR301240-25GEA 242 EUR
APOSOR301240-25G APOSOR301240-5G
5-Bromopyrimidine-2-carboxylic acid 97%
5-Bromopyrimidine-2-carboxylic acid
Formula: C₅H₃BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00496793
CAS Number: 37131-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR