2-Amino-5-bromo-3-pyridinol 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Amino-5-bromo-3-hydroxypyridine
OR301243-25G
OR301243-5G
OR301243-100G
APOSOR301243-25GEA
220
EUR
APOSOR301243-25G
APOSOR301243-5G
APOSOR301243-100G
2-Amino-5-bromo-3-pyridinol 98%
2-Amino-5-bromo-3-pyridinol
Formula:
C₅H₅BrN₂O MW: 189,01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09744143 CAS Number: 39903-01-0 EINECS: 689-734-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...