6-Chloro-1H-pyrrolo[3,2-c]pyridine 97%

Supplier: Apollo Scientific
Warning

Synonyms: 6-Chloro-5-azaindole

OR301258-1G OR301258-5G
APOSOR301258-1GEA 77.1 EUR
APOSOR301258-1G APOSOR301258-5G
6-Chloro-1H-pyrrolo[3,2-c]pyridine 97%
6-Chloro-1H-pyrrolo[3,2-c]pyridine
Formula: C₇H₅ClN₂
MW: 152,5800 g/mol
Melting Pt: 190…191 °C
Storage Temperature: Ambient
MDL Number: MFCD08690133
CAS Number: 74976-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR