Cyclopropyl isothiocyanate 97%

Supplier: Apollo Scientific
Danger

OR30127-1G
APOSOR30127-1GEA 26.1 EUR
APOSOR30127-1G
Cyclopropyl isothiocyanate 97%
Cyclopropyl isothiocyanate
Formula: C₄H₅NS
MW: 99,16 g/mol
Boiling Pt: 53 °C/20mmHg
Density: 1,053 g/cm³ (25 °C)
Flash Pt: 54,4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00040874
CAS Number: 56601-42-4
EINECS: 260-278-6
UN: 3286
ADR: 3+6.1+8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR