4-Chloro-3-iodopyridine
Supplier: Apollo Scientific
|
Danger
|
OR301298-5G
APOSOR301298-5GEA
68.3
EUR
APOSOR301298-5G
4-Chloro-3-iodopyridine
4-Chloro-3-iodopyridine
Formula:
C₅H₃ClIN MW: 239,4415 g/mol Melting Pt: 71…81 °C Storage Temperature: Freezer |
MDL Number:
MFCD00234024 CAS Number: 89167-34-0 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...