6-Bromo-1-indanone
Supplier: Apollo Scientific
|
Warning
|
OR301319-100G
APOSOR301319-100GEA
125
EUR
APOSOR301319-100G
6-Bromo-1-indanone
6-Bromo-1-indanone
Formula:
C₉H₇BrO MW: 211,06 g/mol Melting Pt: 111…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179286 CAS Number: 14548-39-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...