tert-Butyl-3-hydroxycyclobutylcarbamate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
tert-butyl N-(3-hydroxycyclobutyl)carbamate, tert-Butyl (3-hydroxycyclobutyl)carbamate
OR301347-1G
OR301347-250MG
APOSOR301347-1GEA
16.1
EUR
APOSOR301347-1G
APOSOR301347-250MG
tert-Butyl-3-hydroxycyclobutylcarbamate
tert-Butyl-3-hydroxycyclobutylcarbamate
Formula:
C₉H₁₇NO₃ MW: 187,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11501030 CAS Number: 154748-63-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...