Ethyl trimethylacetopyruvate 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Ethyl 5,5-dimethyl-2,4-dioxohexanoate
OR30152-1G
APOSOR30152-1GEA
65.4
EUR
APOSOR30152-1G
Ethyl trimethylacetopyruvate 97%
Ethyl trimethylacetopyruvate
Formula:
C₁₀H₁₆O₄ MW: 200,24 g/mol Boiling Pt: 68…70 °C/0,5mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00052319 CAS Number: 13395-36-3 EINECS: 236-478-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...