7-Methoxy-3,4,5,6-tetrahydro-2H-azepine

Supplier: Apollo Scientific
Warning

OR30218-25G OR30218-5G
APOSOR30218-25GEA 281 EUR
APOSOR30218-25G APOSOR30218-5G
7-Methoxy-3,4,5,6-tetrahydro-2H-azepine
7-Methoxy-3,4,5,6-tetrahydro-2H-azepine
Formula: C₇H₁₃NO
Boiling Pt: 52…53 °C/18mmHg
Density: 0,887 g/cm³ (25 °C)
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00006938
CAS Number: 2525-16-8
EINECS: 219-762-2
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR