4,4-Dimethyl-3,5,8-trioxabicyclo[5.1.0]octane

Supplier: Apollo Scientific
Warning

OR302184-1G OR302184-5G OR302184-10G
APOSOR302184-1GEA 21.9 EUR
APOSOR302184-1G APOSOR302184-5G APOSOR302184-10G
4,4-Dimethyl-3,5,8-trioxabicyclo[5.1.0]octane
4,4-Dimethyl-3,5,8-trioxabicyclo[5.1.0]octane
Formula: C₇H₁₂O₃
MW: 144,17 g/mol
Boiling Pt: 199,8 °C
Melting Pt: –7 °C
Density: 1,11 g/cm³ (25 °C)
Flash Pt: 85,5 °C
Storage Temperature: Ambient
MDL Number: MFCD16621162
CAS Number: 57280-22-5
EINECS: 421-750-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR