Pyridine-2,4-dicarboxylic acid 97%

Supplier: Apollo Scientific
Warning

Synonyms: Lutidinic acid, 2,4-Pyridinedicarboxylic acid, PCA, 2,4-PDCA

OR302207-100G OR302207-5G OR302207-25G
APOSOR302207-100GEA 161 EUR
APOSOR302207-100G APOSOR302207-5G APOSOR302207-25G
Pyridine-2,4-dicarboxylic acid 97%
Pyridine-2,4-dicarboxylic acid
Formula: C₇H₅NO₄
MW: 167,12 g/mol
Melting Pt: 246…248 °C
Storage Temperature: Ambient
MDL Number: MFCD00149413
CAS Number: 499-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR