1,3-Diphenyl-1H-pyrazol-5-amine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Amino-1,3-diphenylpyrazole
OR3023-1G
OR3023-250MG
APOSOR3023-1GEA
32.1
EUR
APOSOR3023-1G
APOSOR3023-250MG
1,3-Diphenyl-1H-pyrazol-5-amine
1,3-Diphenyl-1H-pyrazol-5-amine
Formula:
C₁₅H₁₃N₃ MW: 235,29 g/mol Melting Pt: 129…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00084878 CAS Number: 5356-71-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...