Nortropine

Supplier: Apollo Scientific
Warning

Synonyms: (3-endo)-8-Azabicyclo[3.2.1]octan-3-ol, 8-Azabicyclo[3.2.1]octan-3-ol

OR302770-25G OR302770-5G OR302770-50G
APOSOR302770-25GEA 66.7 EUR
APOSOR302770-25G APOSOR302770-5G APOSOR302770-50G
Nortropine
Nortropine
Formula: C₇H₁₃NO
MW: 127,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047140
CAS Number: 538-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR