2-Amino-N-methyl-2-phenylacetamide
Supplier: Apollo Scientific
|
Warning
|
OR303002-1G
OR303002-500MG
APOSOR303002-1GEA
336
EUR
APOSOR303002-1G
APOSOR303002-500MG
2-Amino-N-methyl-2-phenylacetamide
2-Amino-N-methyl-2-phenylacetamide
Formula:
C₉H₁₂N₂O Storage Temperature: Ambient |
MDL Number:
MFCD09948081 CAS Number: 93782-07-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...