Phosphonitrilic chloride trimer 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
Cyclic Phosphonitrilic Chloride Trimer, Hexachlorocyclotriphosphazene
OR30304-100G
OR30304-25G
OR30304-500G
APOSOR30304-100GEA
84.1
EUR
APOSOR30304-100G
APOSOR30304-25G
APOSOR30304-500G
Phosphonitrilic chloride trimer 97%
Phosphonitrilic chloride trimer
Formula:
Cl₆N₃P₃ Boiling Pt: 127 °C/13mmHg Melting Pt: 111…115 °C Density: 1,98 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006474 CAS Number: 940-71-6 EINECS: 213-376-8 UN: 3260 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...