Benzyl-4-[methoxy(methyl)carbamoyl]piperidine-1-carboxylate
Supplier: Apollo Scientific
OR303056-1G
OR303056-500MG
APOSOR303056-1GEA
210
EUR
APOSOR303056-1G
APOSOR303056-500MG
Benzyl-4-[methoxy(methyl)carbamoyl]piperidine-1-carboxylate
Benzyl-4-[methoxy(methyl)carbamoyl]piperidine-1-carboxylate
Formula:
C₁₆H₂₂N₂O₄ MW: 306,36 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07370044 CAS Number: 148148-48-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...