4-(Benzylsulphanyl)-3-nitrobenzenecarbaldehyde

Supplier: Apollo Scientific
Warning

OR30328-100MG OR30328-250MG
APOSOR30328-100MGEA 21.9 EUR
APOSOR30328-100MG APOSOR30328-250MG
4-(Benzylsulphanyl)-3-nitrobenzenecarbaldehyde
4-(Benzylsulphanyl)-3-nitrobenzenecarbaldehyde
Formula: C₁₄H₁₁NO₃S
Melting Pt: 170…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00173710
CAS Number: 175278-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR