3-cyclopropyl-1H-pyrazole 97+%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Cyclopropyl-1H-pyrazole
OR303295-5G
APOSOR303295-5GEA
79.9
EUR
APOSOR303295-5G
3-cyclopropyl-1H-pyrazole 97+%
3-cyclopropyl-1H-pyrazole
Formula:
C₆H₈N₂ MW: 108,144 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD0497093 CAS Number: 100114-57-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...