Ethyl potassium oxalate
Supplier: Apollo Scientific
|
Warning
|
OR303345-25G
OR303345-5G
APOSOR303345-25GEA
145
EUR
APOSOR303345-25G
APOSOR303345-5G
Ethyl potassium oxalate
Ethyl potassium oxalate
Formula:
C₄H₅KO₄ MW: 156,178 g/mol Melting Pt: 220…223 °C Storage Temperature: Ambient |
MDL Number:
MFCD00045861 CAS Number: 1906-57-6 EINECS: 217-606-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...