4-Bromo-4'-chloro-1,1'-biphenyl
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Bromo-4'-Chloro-1,1'-biphenyl
OR303367-1G
OR303367-5G
APOSOR303367-1GEA
53.9
EUR
APOSOR303367-1G
APOSOR303367-5G
4-Bromo-4'-chloro-1,1'-biphenyl
4-Bromo-4'-chloro-1,1'-biphenyl
Formula:
C₁₂H₈BrCl MW: 267,55 g/mol Melting Pt: 150…153 °C Storage Temperature: Ambient |
MDL Number:
MFCD06201384 CAS Number: 23055-77-8 UN: 3152 ADR: 9,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...