5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxaldehyde
Supplier: Apollo Scientific
|
Warning
|
OR303422-25G
OR303422-5G
OR303422-1G
APOSOR303422-25GEA
68.7
EUR
APOSOR303422-25G
APOSOR303422-5G
APOSOR303422-1G
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxaldehyde
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxaldehyde
Formula:
C₁₁H₁₅BO₃S Melting Pt: 73 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09266186 CAS Number: 1040281-83-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...