3-Iodophenylboronic acid

Supplier: Apollo Scientific

OR303431-1G OR303431-5G
APOSOR303431-1GEA 0 EUR
APOSOR303431-1G APOSOR303431-5G
3-Iodophenylboronic acid
3-Iodophenylboronic acid
Formula: C₆H₆BIO₂
Storage Temperature: Ambient
MDL Number: MFCD01319015
CAS Number: 221037-98-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR