9,9-Dimethylfluorene-2-boronic acid pinacol ester

Supplier: Apollo Scientific

Synonyms: 9,9-Dimethylfluorene-2-boronic acid Pinacol ester, 9,9-Dimethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl)-9H-fluorene, 2-(9,9-Dimethyl-9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR303498-10G OR303498-1G OR303498-5G
APOSOR303498-10GEA 136 EUR
APOSOR303498-10G APOSOR303498-1G APOSOR303498-5G
9,9-Dimethylfluorene-2-boronic acid pinacol ester
9,9-Dimethylfluorene-2-boronic acid pinacol ester
Formula: C₂₁H₂₅BO₂
MW: 320,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08704229
CAS Number: 569343-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR